2-[5-(Trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid
Supplier: Apollo Scientific
Warning
PC8059-1G PC8059-10G PC8059-5G
APOSPC8059-1GEA 248 EUR
APOSPC8059-1G APOSPC8059-10G APOSPC8059-5G
2-[5-(Trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid
2-[5-(Trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid
Formula: C₈H₄F₃N₃O₂S
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD09836179
CAS Number: 1209904-67-5