2-Bromo-3-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Warning
PC8075-5G PC8075-1G PC8075-25G
APOSPC8075-5GEA 29 EUR
APOSPC8075-5G APOSPC8075-1G APOSPC8075-25G
2-Bromo-3-(trifluoromethyl)aniline 98%
2-Bromo-3-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01656033
CAS Number: 58458-10-9