2-Amino-6-fluorobenzothiazole 99%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Fluorobenzothiazol-2-ylamine, 6-Fluorobenzo[d]thiazol-2-amine, 6-Fluoro-1,3-benzothiazol-2-amine
PC8076-25G PC8076-5G PC8076-1G PC8076-100G
APOSPC8076-25GEA 94.4 EUR
APOSPC8076-25G APOSPC8076-5G APOSPC8076-1G APOSPC8076-100G
2-Amino-6-fluorobenzothiazole 99%
2-Amino-6-fluorobenzothiazole
Formula: C₇H₅FN₂S
MW: 168.19 g/mol
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00013336
CAS Number: 348-40-3
EINECS: 609-034-6