5-(2-Chlorobenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde 97%
Supplier: Apollo Scientific
PC8130-1G
PC8130-500MG
PC8130-5G
APOSPC8130-1GEA
533
EUR
APOSPC8130-1G
APOSPC8130-500MG
APOSPC8130-5G
5-(2-Chlorobenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde 97%
5-(2-Chlorobenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde
Formula:
C₁₃H₁₀ClF₃N₂OS Melting Pt: 86…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00140828 CAS Number: 321533-90-8 |