2'-Fluoro-4'-hydroxyacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2-Fluoro-4-hydroxyphenyl)ethanone
PC8139-100G PC8139-25G PC8139-5G
APOSPC8139-100GEA 214 EUR
APOSPC8139-100G APOSPC8139-25G APOSPC8139-5G
2'-Fluoro-4'-hydroxyacetophenone
2'-Fluoro-4'-hydroxyacetophenone
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 121…123 °C
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00142713
CAS Number: 98619-07-9