2-Chloro-4-fluorocinnamic acid 98%
Supplier: Apollo Scientific
Warning
PC8152-1G PC8152-25G PC8152-5G
APOSPC8152-1GEA 40.8 EUR
APOSPC8152-1G APOSPC8152-25G APOSPC8152-5G
2-Chloro-4-fluorocinnamic acid 98%
2-Chloro-4-fluorocinnamic acid
Formula: C₉H₆ClFO₂
MW: 200.6 g/mol
Melting Pt: 247…249 °C
Storage Temperature: Ambient
MDL Number: MFCD00236027
CAS Number: 133220-86-7