1-Bromo-2,5-difluoro-4-nitrobenzene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Brom-2,5-difluor-4-nitrobenzol, 4-Bromo-2,5-difluoronitrobenzene
PC8153-1G
PC8153-5G
APOSPC8153-1GEA
21.9
EUR
APOSPC8153-1G
APOSPC8153-5G
1-Bromo-2,5-difluoro-4-nitrobenzene
1-Bromo-2,5-difluoro-4-nitrobenzene
Formula:
C₆H₂BrF₂NO₂ Melting Pt: 55…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042184 CAS Number: 167415-27-2 |