6,7-Dichloro-2-methyl-3-(trifluoromethyl)quinoxaline 97%
Supplier: Apollo Scientific
Warning
PC8174-250MG PC8174-100MG
APOSPC8174-250MGEA 284 EUR
APOSPC8174-250MG APOSPC8174-100MG
6,7-Dichloro-2-methyl-3-(trifluoromethyl)quinoxaline 97%
6,7-Dichloro-2-methyl-3-(trifluoromethyl)quinoxaline
Formula: C₁₀H₅Cl₂F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD09801054
CAS Number: 143309-87-9