2,6-Dichloro-4-fluoroaniline 97%
Supplier: Apollo Scientific
Danger
PC8181-100G PC8181-25G PC8181-5G
APOSPC8181-100GEA 207 EUR
APOSPC8181-100G APOSPC8181-25G APOSPC8181-5G
2,6-Dichloro-4-fluoroaniline 97%
2,6-Dichloro-4-fluoroaniline
Formula: C₆H₄Cl₂FN
MW: 180.01 g/mol
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00142845
CAS Number: 344-19-4
UN: 2811
ADR: 6.1,III