1,2-Difluoro-4,5-dimethoxybenzene
Supplier: Apollo Scientific
Warning
PC8185-1G PC8185-25G PC8185-5G
APOSPC8185-1GEA 21.9 EUR
APOSPC8185-1G APOSPC8185-25G APOSPC8185-5G
1,2-Difluoro-4,5-dimethoxybenzene
1,2-Difluoro-4,5-dimethoxybenzene
Formula: C₈H₈F₂O₂
MW: 174.15 g/mol
Melting Pt: 41…43 °C
Density: 1.226 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00070789
CAS Number: 203059-80-7