N-(4,5-Difluoro-2-nitrophenyl)acetamide
Supplier: Apollo Scientific
Warning
Synonyms: 4',5'-Difluoro-2'-nitroacetanilide
PC8186-1G PC8186-5G
APOSPC8186-1GEA 50.9 EUR
APOSPC8186-1G APOSPC8186-5G
N-(4,5-Difluoro-2-nitrophenyl)acetamide
N-(4,5-Difluoro-2-nitrophenyl)acetamide
Formula: C₈H₆F₂N₂O₃
MW: 216.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143199
CAS Number: 1662-21-1