2-Fluoro-4-methoxybenzoic acid
Supplier: Apollo Scientific
Warning
PC8207-25G PC8207-5G PC8207-100G
APOSPC8207-25GEA 97.3 EUR
APOSPC8207-25G APOSPC8207-5G APOSPC8207-100G
2-Fluoro-4-methoxybenzoic acid
2-Fluoro-4-methoxybenzoic acid
Formula: C₈H₇FO₃
MW: 170.14 g/mol
Melting Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00509373
CAS Number: 394-42-3