2-Fluoro-4-methoxyphenol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Fluoro-4-hydroxyanisole
PC8211-5G PC8211-25G PC8211-50G PC8211-100G
APOSPC8211-5GEA 50.9 EUR
APOSPC8211-5G APOSPC8211-25G APOSPC8211-50G APOSPC8211-100G
2-Fluoro-4-methoxyphenol 97%
2-Fluoro-4-methoxyphenol
Formula: C₇H₇FO₂
MW: 142.13 g/mol
Boiling Pt: 94 °C/3.75mmHg
Density: 1.27 g/cm³ (25 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00070796
CAS Number: 167683-93-4