3-Fluoro-4-methylanisole 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-4-methoxy-1-methylbenzene
PC8219-25G PC8219-1G PC8219-5G
APOSPC8219-25GEA 119 EUR
APOSPC8219-25G APOSPC8219-1G APOSPC8219-5G
3-Fluoro-4-methylanisole 99%
3-Fluoro-4-methylanisole
Formula: C₈H₉FO
MW: 140.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143163
CAS Number: 405-06-1