1,2-Dichloro-1,1,2-trifluoro-2-(trifluoromethoxy)ethane 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,2-Dichloro-1,2,2-trifluoroethyl trifluoromethyl ether
PC8222-25G PC8222-5G
APOSPC8222-25GEA 263 EUR
APOSPC8222-25G APOSPC8222-5G
1,2-Dichloro-1,1,2-trifluoro-2-(trifluoromethoxy)ethane 98%
1,2-Dichloro-1,1,2-trifluoro-2-(trifluoromethoxy)ethane
Formula: ClF₂CCClFOCF₃
MW: 236.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464450
CAS Number: 2356-53-8