4-(Trifluoromethyl)anisole
Supplier: Apollo Scientific
|
Warning
|
PC8229-5G
PC8229-1G
PC8229-25G
APOSPC8229-5GEA
31.9
EUR
APOSPC8229-5G
APOSPC8229-1G
APOSPC8229-25G
4-(Trifluoromethyl)anisole
4-(Trifluoromethyl)anisole
Formula:
C₈H₇F₃O MW: 176.14 g/mol Boiling Pt: 166…170 °C Melting Pt: –9 °C Density: 1.2445 g/cm³ (25 °C) Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094456 CAS Number: 402-52-8 UN: 1993 ADR: 3 III |