5-(Trifluoromethyl)thieno[3,2-b]pyridine-6-carboxylic acid 95+%
Supplier: Apollo Scientific
Warning
PC8252-1G PC8252-250MG
APOSPC8252-1GEA 1850 EUR
APOSPC8252-1G APOSPC8252-250MG
5-(Trifluoromethyl)thieno[3,2-b]pyridine-6-carboxylic acid 95+%
5-(Trifluoromethyl)thieno[3,2-b]pyridine-6-carboxylic acid
Formula: C₉H₄F₃NO₂S
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00203137
CAS Number: 243977-22-2