2,6-Difluoro-4-hydroxybenzonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Cyano-3,5-difluorophenol
PC8318-1G PC8318-25G PC8318-5G
APOSPC8318-1GEA 58.2 EUR
APOSPC8318-1G APOSPC8318-25G APOSPC8318-5G
2,6-Difluoro-4-hydroxybenzonitrile 98%
2,6-Difluoro-4-hydroxybenzonitrile
Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD03094499
CAS Number: 123843-57-2
UN: 3439
ADR: 6.1,III