2-Chloro-4-(trifluoromethoxy)aniline
Supplier: Apollo Scientific
Danger
PC8324-25G PC8324-5G
APOSPC8324-25GEA 79.9 EUR
APOSPC8324-25G APOSPC8324-5G
2-Chloro-4-(trifluoromethoxy)aniline
2-Chloro-4-(trifluoromethoxy)aniline
Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
Boiling Pt: 94…96 °C/11mmHg
Density: 1.524 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD01631704
CAS Number: 69695-61-0