Fmoc-2-(trifluoromethyl)-D-phenylalanine
Supplier: Apollo Scientific
Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid, 2-(Trifluoromethyl)-D-phenylalanine, N-FMOC protected, Fmoc-D-Phe(2-CF₃)-OH
PC8357-1G PC8357-250MG
APOSPC8357-1GEA 48 EUR
APOSPC8357-1G APOSPC8357-250MG
Fmoc-2-(trifluoromethyl)-D-phenylalanine
Fmoc-2-(trifluoromethyl)-D-phenylalanine
Formula: C₂₅H₂₀F₃NO₄
MW: 455.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01863051
CAS Number: 352523-15-0