4,5-Dichloro-2-[4-(trifluoromethyl)phenyl]pyridazin-3(2H)-one 95%
Supplier: Apollo Scientific
Warning
PC8377-100MG PC8377-1G PC8377-250MG
APOSPC8377-100MGEA 31.9 EUR
APOSPC8377-100MG APOSPC8377-1G APOSPC8377-250MG
4,5-Dichloro-2-[4-(trifluoromethyl)phenyl]pyridazin-3(2H)-one 95%
4,5-Dichloro-2-[4-(trifluoromethyl)phenyl]pyridazin-3(2H)-one
Formula: C₁₁H₅Cl₂F₃N₂O
MW: 309.08 g/mol
Melting Pt: 139…140 °C
Storage Temperature: Ambient
MDL Number: MFCD01313341
CAS Number: 62436-07-1