3-Chloro-5-fluoropyridine
Supplier: Apollo Scientific
Danger
PC8386-1G PC8386-5G
APOSPC8386-1GEA 21.9 EUR
APOSPC8386-1G APOSPC8386-5G
3-Chloro-5-fluoropyridine
3-Chloro-5-fluoropyridine
Formula: C₅H₃ClFN
MW: 131.54 g/mol
Melting Pt: 25…30 °C
Flash Pt: 41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663707
CAS Number: 514797-99-0
UN: 1325
ADR: 4.1 III