6-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine
Supplier: Apollo Scientific
Synonyms:
2-Amino-6-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine
PC8436-1G
PC8436-500MG
PC8436-5G
APOSPC8436-1GEA
799
EUR
APOSPC8436-1G
APOSPC8436-500MG
APOSPC8436-5G
6-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine
6-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine
Formula:
C₁₃H₁₁ClF₃N₅ MW: 329.71 g/mol Melting Pt: 200…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD00202227 CAS Number: 338979-12-7 |