2-[Cyano(2,4-dichlorophenyl)methyl]-6-fluorobenzenecarbonitrile 97%
Supplier: Apollo Scientific
Synonyms: 2-[Cyano(2,4-dichlorophenyl)methyl]-6-fluorobenzonitrile
PC8438-10G PC8438-1G PC8438-5G
APOSPC8438-10GEA 379 EUR
APOSPC8438-10G APOSPC8438-1G APOSPC8438-5G
2-[Cyano(2,4-dichlorophenyl)methyl]-6-fluorobenzenecarbonitrile 97%
2-[Cyano(2,4-dichlorophenyl)methyl]-6-fluorobenzenecarbonitrile
Formula: C₁₅H₇Cl₂FN₂
MW: 305.14 g/mol
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00231988
CAS Number: 338965-45-0