2,3-Diamino-6-(trifluoromethyl)-4-(3H)-pyrimidinone 97%
Supplier: Apollo Scientific
Warning
PC8450-1G PC8450-500MG
APOSPC8450-1GEA 34.8 EUR
APOSPC8450-1G APOSPC8450-500MG
2,3-Diamino-6-(trifluoromethyl)-4-(3H)-pyrimidinone 97%
2,3-Diamino-6-(trifluoromethyl)-4-(3H)-pyrimidinone
Formula: C₅H₅F₃N₄O
MW: 194.12 g/mol
Melting Pt: 217…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00068010
CAS Number: 95095-71-9