3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]acrylonitrile 97%
Supplier: Apollo Scientific
PC8458-1G PC8458-500MG PC8458-5G
APOSPC8458-1GEA 172 EUR
APOSPC8458-1G APOSPC8458-500MG APOSPC8458-5G
3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]acrylonitrile 97%
3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]acrylonitrile
Formula: C₁₂H₁₁F₃N₂
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00215017
CAS Number: 62738-99-2