1,3-Dimethyl-5[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carboxylic acid 97%
Supplier: Apollo Scientific
PC8460-1G PC8460-500MG
APOSPC8460-1GEA 666 EUR
APOSPC8460-1G APOSPC8460-500MG
1,3-Dimethyl-5[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carboxylic acid 97%
1,3-Dimethyl-5[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carboxylic acid
Formula: C₁₃H₁₁F₃N₂O₃
MW: 300.24 g/mol
Melting Pt: 200…203 °C
Storage Temperature: Ambient
MDL Number: MFCD01935857
CAS Number: 446276-23-9