1-[2,6-Dinitro-4-(trifluoromethyl)phenyl]piperazine 97%
Supplier: Apollo Scientific
Warning
PC8462-1G PC8462-250MG PC8462-5G
APOSPC8462-1GEA 165 EUR
APOSPC8462-1G APOSPC8462-250MG APOSPC8462-5G
1-[2,6-Dinitro-4-(trifluoromethyl)phenyl]piperazine 97%
1-[2,6-Dinitro-4-(trifluoromethyl)phenyl]piperazine
Formula: C₁₁H₁₁F₃N₄O₄
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00052948
CAS Number: 36317-84-7