3,5-Dioxo-2-[3-(trifluoromethyl)phenyl]-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile 97%
Supplier: Apollo Scientific
PC8464-500MG PC8464-5G PC8464-1G
APOSPC8464-500MGEA 302 EUR
APOSPC8464-500MG APOSPC8464-5G APOSPC8464-1G
3,5-Dioxo-2-[3-(trifluoromethyl)phenyl]-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile 97%
3,5-Dioxo-2-[3-(trifluoromethyl)phenyl]-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile
Formula: C₁₁H₅F₃N₄O₂
MW: 282.18 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00274940
CAS Number: 121065-97-2