5-Bromo-2,3-difluoroanisole 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-1,2-difluoro-3-methoxybenzene
PC8472-100G
APOSPC8472-100GEA 155 EUR
APOSPC8472-100G
5-Bromo-2,3-difluoroanisole 97%
5-Bromo-2,3-difluoroanisole
Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD01631352
CAS Number: 261762-35-0