Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}acetate 95%
Supplier: Apollo Scientific
Synonyms: Ethyl N-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]glycinate
PC8474-10G PC8474-1G PC8474-5G
APOSPC8474-10GEA 619 EUR
APOSPC8474-10G APOSPC8474-1G APOSPC8474-5G
Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}acetate 95%
Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}acetate
Formula: C₁₀H₁₀ClF₃N₂O₂
MW: 282.65 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00141296
CAS Number: 246022-36-6