4-Methoxy-3-(trifluoromethyl)benzoic acid
Supplier: Apollo Scientific
Warning
PC8475-25G PC8475-5G PC8475-100G
APOSPC8475-25GEA 52.2 EUR
APOSPC8475-25G APOSPC8475-5G APOSPC8475-100G
4-Methoxy-3-(trifluoromethyl)benzoic acid
4-Methoxy-3-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD01091014
CAS Number: 213598-09-5