2-Bromo-N-(4-(trifluoromethyl)phenyl)acetamide
Supplier: Apollo Scientific
Warning
Synonyms: 4-[(Bromoacetyl)amino]benzotrifluoride, N-(Bromoacetyl)-4-(trifluoromethyl)aniline
PC8478-25G PC8478-5G PC8478-1G
APOSPC8478-25GEA 636 EUR
APOSPC8478-25G APOSPC8478-5G APOSPC8478-1G
2-Bromo-N-(4-(trifluoromethyl)phenyl)acetamide
2-Bromo-N-(4-(trifluoromethyl)phenyl)acetamide
Formula: C₉H₇BrF₃NO
MW: 282.06 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00662747
CAS Number: 3823-19-6