3-Chloro-4-(trifluoromethoxy)benzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
PC8493-1G PC8493-5G PC8493-25G
APOSPC8493-1GEA 58 EUR
APOSPC8493-1G APOSPC8493-5G APOSPC8493-25G
3-Chloro-4-(trifluoromethoxy)benzenesulphonyl chloride 98%
3-Chloro-4-(trifluoromethoxy)benzenesulphonyl chloride
Formula: C₇H₃Cl₂F₃O₃S
MW: 295.07 g/mol
Boiling Pt: 80…84 °C/1mmHg
Density: 1.597 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD04972750
CAS Number: 886762-48-7
UN: 3265
ADR: 8 III