2-(Trifluoromethyl)benzenecarbaldehyde oxime
Supplier: Apollo Scientific
PC8499-1G
PC8499-5G
APOSPC8499-1GEA
72.5
EUR
APOSPC8499-1G
APOSPC8499-5G
2-(Trifluoromethyl)benzenecarbaldehyde oxime
2-(Trifluoromethyl)benzenecarbaldehyde oxime
Formula:
C₈H₆F₃NO Melting Pt: 55…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD03015196 CAS Number: 74467-00-8 |