4-Bromo-3-fluorophenol 97%
Supplier: Apollo Scientific
Warning
PC8537-100G PC8537-5G PC8537-25G
APOSPC8537-100GEA 0 EUR
APOSPC8537-100G APOSPC8537-5G APOSPC8537-25G
4-Bromo-3-fluorophenol 97%
4-Bromo-3-fluorophenol
Formula: C₆H₄BrFO
Boiling Pt: 96 °C/2.8mmHg
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00051907
CAS Number: 121219-03-2