3,5-Difluorobenzhydrazide 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Difluorobenzohydrazide, 3,5-Difluorobenzoic hydrazide, 3,5-Difluorobenzene-1-carbohydrazide, 3,5-Difluorobenzoic acid hydrazide
PC8562-1G PC8562-5G PC8562-10G
APOSPC8562-1GEA 33.5 EUR
APOSPC8562-1G APOSPC8562-5G APOSPC8562-10G
3,5-Difluorobenzhydrazide 97%
3,5-Difluorobenzhydrazide
Formula: C₇H₆F₂N₂O
MW: 172.13 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01310810
CAS Number: 244022-63-7