2-Fluoro-5-iodobenzoic acid
Supplier: Apollo Scientific
Warning
PC8563-1G PC8563-25G PC8563-5G PC8563-100G
APOSPC8563-1GEA 14.7 EUR
APOSPC8563-1G APOSPC8563-25G APOSPC8563-5G APOSPC8563-100G
2-Fluoro-5-iodobenzoic acid
2-Fluoro-5-iodobenzoic acid
Formula: C₇H₄FIO₂
MW: 266.01 g/mol
Melting Pt: 162…168 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094517
CAS Number: 124700-41-0