5-Bromo-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Supplier: Apollo Scientific
PC8570-1G PC8570-500MG
APOSPC8570-1GEA 346 EUR
APOSPC8570-1G APOSPC8570-500MG
5-Bromo-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
5-Bromo-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Formula: C₁₃H₈BrClFNO₂
MW: 344.57 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD09817442
CAS Number: 952183-68-5