3-(Trifluoromethyl)thiobenzamide 95+%
Supplier: Apollo Scientific
Warning
PC8573-5G PC8573-25G
APOSPC8573-5GEA 168 EUR
APOSPC8573-5G APOSPC8573-25G
3-(Trifluoromethyl)thiobenzamide 95+%
3-(Trifluoromethyl)thiobenzamide
Formula: C₈H₆F₃NS
Melting Pt: 69…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01567258
CAS Number: 53515-17-6
UN: 2811
ADR: 6.1,III