2-Bromo-5-fluorobenzonitrile 99%
Supplier: Apollo Scientific
Warning
PC8576-100G PC8576-25G PC8576-5G
APOSPC8576-100GEA 108 EUR
APOSPC8576-100G APOSPC8576-25G APOSPC8576-5G
2-Bromo-5-fluorobenzonitrile 99%
2-Bromo-5-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 255
Melting Pt: 88…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00142875
CAS Number: 57381-39-2
EINECS: 260-711-9
UN: 3439
ADR: 6.1 III