3-Bromo-2-fluorobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC8581-25G PC8581-5G PC8581-100G
APOSPC8581-25GEA 49.3 EUR
APOSPC8581-25G APOSPC8581-5G APOSPC8581-100G
3-Bromo-2-fluorobenzaldehyde 98%
3-Bromo-2-fluorobenzaldehyde
Formula: C₇H₄BrFO
Melting Pt: 41…46 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD09027089
CAS Number: 149947-15-9