Ethyl 2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole-5-carboxylate
Supplier: Apollo Scientific
Warning
PC8585-1G PC8585-5G
APOSPC8585-1GEA 263 EUR
APOSPC8585-1G APOSPC8585-5G
Ethyl 2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole-5-carboxylate
Ethyl-2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole-5-carboxylate
Formula: C₁₄H₁₁F₄NO₂S
MW: 333.31 g/mol
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD07369386
CAS Number: 317319-21-4