Methyl 5-chloro-6-oxo-1-[4-(trifluoromethyl)-benzyl]-1,6-dihydro-3-pyridinecarboxylate
Supplier: Apollo Scientific
PC8594-10G PC8594-1G PC8594-5G
APOSPC8594-10GEA 477 EUR
APOSPC8594-10G APOSPC8594-1G APOSPC8594-5G
Methyl 5-chloro-6-oxo-1-[4-(trifluoromethyl)-benzyl]-1,6-dihydro-3-pyridinecarboxylate
Methyl-5-chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridinecarboxylate
Formula: C₁₅H₁₁ClF₃NO₃
MW: 345.71 g/mol
Melting Pt: 169…170 °C
Storage Temperature: Ambient
MDL Number: MFCD09027141
CAS Number: 937602-45-4