Methyl-6-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Supplier: Apollo Scientific
Warning
PC8612-1G PC8612-5G
APOSPC8612-1GEA 163 EUR
APOSPC8612-1G APOSPC8612-5G
Methyl-6-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Methyl 6-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Formula: C₁₁H₇F₃O₂S
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD07371545
CAS Number: 863118-41-6