Methyl-2,5-difluoro-4-nitrobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2,5-difluoro-4-nitrobenzoate
PC8637-1G PC8637-5G PC8637-10G
APOSPC8637-1GEA 21.9 EUR
APOSPC8637-1G APOSPC8637-5G APOSPC8637-10G
Methyl-2,5-difluoro-4-nitrobenzoate
Methyl 2,5-difluoro-4-nitrobenzoate
Formula: C₈H₅F₂NO₄
MW: 217.13 g/mol
Melting Pt: 49…52 °C
Storage Temperature: Ambient
MDL Number: MFCD08689670
CAS Number: 924868-81-5