6-Fluoro-m-toluonitrile 98%
Supplier: Apollo Scientific
Danger
PC8655-1G PC8655-25G PC8655-5G
APOSPC8655-1GEA 21.9 EUR
APOSPC8655-1G APOSPC8655-25G APOSPC8655-5G
6-Fluoro-m-toluonitrile 98%
6-Fluoro-m-toluonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 204 °C
Melting Pt: 50…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00134542
CAS Number: 64113-84-4
UN: 2811
ADR: 6.1 III