2-Chloro-3,5-difluoroaniline 97%
Supplier: Apollo Scientific
Warning
PC8712-5G PC8712-25G PC8712-50G PC8712-100G
APOSPC8712-5GEA 46.4 EUR
APOSPC8712-5G APOSPC8712-25G APOSPC8712-50G APOSPC8712-100G
2-Chloro-3,5-difluoroaniline 97%
2-Chloro-3,5-difluoroaniline
Formula: C₆H₄ClF₂N
MW: 163.5525 g/mol
Melting Pt: 26…28 °C
Density: 1.434 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134427
CAS Number: 36556-60-2
UN: 2811
ADR: 6.1,III