3-{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-indol-3-yl}acrylic acid 97%
Supplier: Apollo Scientific
PC8713-1G PC8713-500MG
APOSPC8713-1GEA 799 EUR
APOSPC8713-1G APOSPC8713-500MG
3-{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-indol-3-yl}acrylic acid 97%
3-{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-indol-3-yl}acrylic acid
Formula: C₁₇H₁₀ClF₃N₂O₂
MW: 366.73 g/mol
Melting Pt: 250…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00664519
CAS Number: 1164457-90-2