4-(Trifluoromethyl)styrene 96%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Trifluoromethyl)-4-vinylbenzene
PC8732-1G PC8732-5G
APOSPC8732-1GEA 30.6 EUR
APOSPC8732-1G APOSPC8732-5G
4-(Trifluoromethyl)styrene 96%
4-(Trifluoromethyl)styrene
Formula: C₉H₇F₃
MW: 172.15 g/mol
Boiling Pt: 65…66 °C/40mmHg
Density: 1.165 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075539
CAS Number: 402-50-6
UN: 1993
ADR: 3 III